The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003366.3 | NP_003357.2 | 10 - 11 | 1321 | 91 |
GenBank mRNA | AK093106.1 | - | 5 - 6 | 1141 | 91 |
AK094006.1 | - | 10 - 11 | 1725 | 91 | |
BC000484.2 | - | 10 - 11 | 1044 | 91 | |
BC003136.1 | - | 10 - 11 | 1012 | 91 | |
J04973.1 | - | 10 - 11 | 1014 | 91 |