The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001407.2 | NP_001398.2 | 20 - 21 | 7407 | 137 |
GenBank mRNA | AB011536.1 | - | 10 - 11 | 1283 | 137 |
AF231023.1 | - | 20 - 21 | 7407 | 137 |