The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001289112.1 | NP_001276041.1 | 14 - 15 | 1551 | 84 |
NM_130808.2 | NP_570720.1 | 14 - 15 | 2179 | 84 | |
NM_153429.1 | NP_702907.1 | 15 - 16 | 1539 | 84 | |
XM_011512406.2 | XP_011510708.1 | 13 - 14 | 1326 | 84 | |
XM_011512407.2 | XP_011510709.1 | 13 - 14 | 1583 | 84 | |
XM_011512408.2 | XP_011510710.1 | 13 - 14 | 2711 | 84 | |
XM_017005693.1 | XP_016861182.1 | 17 - 18 | 2932 | 84 | |
XM_017005695.1 | XP_016861184.1 | 16 - 17 | 1802 | 84 | |
XM_017005696.1 | XP_016861185.1 | 8 - 9 | 696 | 84 | |
GenBank mRNA | AF465770.1 | - | 17 - 18 | 1879 | 84 |
AF465771.1 | - | 15 - 16 | 1539 | 84 | |
AK056175.1 | - | 14 - 15 | 1651 | 84 | |
AK128117.1 | - | 18 - 19 | 3429 | 84 | |
AK295557.1 | - | 14 - 15 | 1466 | 84 | |
BC014396.1 | - | 14 - 15 | 1737 | 84 | |
BC075046.2 | - | 14 - 15 | 1328 | 84 |