The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003366.3 | NP_003357.2 | 4 - 5 | 692 | 75 |
GenBank mRNA | AK094006.1 | - | 4 - 5 | 1096 | 75 |
AK307535.1 | - | 4 - 5 | 458 | 75 | |
BC000484.2 | - | 4 - 5 | 415 | 75 | |
BC003136.1 | - | 4 - 5 | 383 | 75 | |
J04973.1 | - | 4 - 5 | 385 | 75 |