Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RUNX1T1
Entrez Gene ID
862
Gene Name
RUNX1 translocation partner 1
Gene Aliases
AML1-MTG8, AML1T1, CBFA2T1, CDR, ETO, MTG8, ZMYND2
Chromosome Location
Chr.8: 91954967 - 92103365 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001198625.1 NP_001185554.1 12 - 13 1962 57
NM_001198626.1 NP_001185555.1 10 - 11 1820 57
NM_001198627.1 NP_001185556.1 11 - 12 1880 57
NM_001198628.1 NP_001185557.1 9 - 10 1477 57
NM_001198629.1 NP_001185558.1 9 - 10 1603 57
NM_001198630.1 NP_001185559.1 10 - 11 1663 57
NM_001198631.1 NP_001185560.1 9 - 10 1676 57
NM_001198632.1 NP_001185561.1 9 - 10 1594 57
NM_001198633.1 NP_001185562.1 8 - 9 1565 57
NM_001198634.1 NP_001185563.1 8 - 9 1503 57
NM_001198679.1 NP_001185608.1 9 - 10 1647 57
NM_004349.3 NP_004340.1 8 - 9 1613 57
NM_175634.2 NP_783552.1 9 - 10 1738 57
NM_175635.2 NP_783553.1 8 - 9 1430 57
NM_175636.2 NP_783554.1 7 - 8 1512 57
XM_006716676.3 XP_006716739.1 10 - 11 1547 57
XM_011517351.2 XP_011515653.1 10 - 11 1592 57
XM_011517352.2 XP_011515654.1 10 - 11 1832 57
XM_011517353.2 XP_011515655.1 9 - 10 1540 57
XM_017013930.1 XP_016869419.1 9 - 10 1848 57
XM_017013931.1 XP_016869420.1 8 - 9 1370 57
XM_017013932.1 XP_016869421.1 10 - 11 1735 57
XM_017013933.1 XP_016869422.1 11 - 12 1799 57
XM_017013934.1 XP_016869423.1 11 - 12 1907 57
XM_017013935.1 XP_016869424.1 10 - 11 1712 57
XM_017013936.1 XP_016869425.1 9 - 10 1806 57
XM_017013937.1 XP_016869426.1 10 - 11 1632 57
XM_017013939.1 XP_016869428.1 4 - 5 1263 57
XM_017013940.1 XP_016869429.1 5 - 6 2492 57
XM_017013941.1 XP_016869430.1 4 - 5 2282 57
GenBank mRNA AK297616.1 - 8 - 9 1327 57
AK312592.1 - 12 - 13 1724 57
BC005850.1 - 8 - 9 1392 57
BC067078.1 - 8 - 9 1383 57
BT009871.1 - 7 - 8 1172 57
CR456792.1 - 7 - 8 1202 57
D14289.1 - 7 - 8 1469 57
D14821.1 - 8 - 9 1367 57
D43638.1 - 8 - 9 1613 57
KF946508.1 - 8 - 9 1231 57
KF946514.1 - 8 - 9 1217 57
KF946517.1 - 7 - 8 1457 57
KF946518.1 - 7 - 8 1457 57
KF946521.1 - 4 - 5 879 57
KF946523.1 - 4 - 5 879 57
X79990.1 - 7 - 8 1195 57

View Full Product Details