The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001316329.1 | NP_001303258.1 | 15 - 16 | 1970 | 91 |
NM_002739.4 | NP_002730.1 | 15 - 16 | 1970 | 91 | |
GenBank mRNA | AF345987.1 | - | 15 - 16 | 1854 | 91 |
AK296412.1 | - | 15 - 16 | 1978 | 91 | |
AK296434.1 | - | 15 - 16 | 1957 | 91 | |
AK312302.1 | - | 15 - 16 | 1932 | 91 | |
BC047876.1 | - | 15 - 16 | 1845 | 91 | |
Z15114.1 | - | 11 - 12 | 1178 | 91 |