The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001145939.1 | NP_001139411.1 | 2 - 3 | 432 | 82 |
NM_001145940.1 | NP_001139412.1 | 2 - 3 | 193 | 82 | |
NM_002573.3 | NP_002564.1 | 1 - 2 | 410 | 82 | |
XM_017026846.1 | XP_016882335.1 | 1 - 2 | 262 | 82 | |
XM_017026847.1 | XP_016882336.1 | 2 - 3 | 427 | 82 | |
XM_017026848.1 | XP_016882337.1 | 2 - 3 | 193 | 82 | |
GenBank mRNA | BC003016.1 | - | 1 - 2 | 337 | 82 |
BC007863.2 | - | 1 - 2 | 208 | 82 | |
CB216829.1 | - | 1 - 2 | 410 | 82 | |
CR407626.1 | - | 1 - 2 | 75 | 82 | |
D63391.1 | - | 1 - 2 | 188 | 82 |