The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006206.4 | NP_006197.1 | 16 - 17 | 2649 | 131 |
XM_005265743.1 | XP_005265800.1 | 16 - 17 | 2433 | 131 | |
XM_006714039.3 | XP_006714102.1 | 17 - 18 | 2694 | 131 | |
XM_011534385.2 | XP_011532687.1 | 16 - 17 | 2687 | 131 | |
XM_017008280.1 | XP_016863769.1 | 16 - 17 | 2864 | 131 | |
GenBank mRNA | AK316578.1 | - | 16 - 17 | 2451 | 131 |
M21574.1 | - | 16 - 17 | 2457 | 131 | |
M22734.1 | - | - | 2446 | 131 |