The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006206.4 | NP_006197.1 | 17 - 18 | 2773 | 65 |
XM_005265743.1 | XP_005265800.1 | 17 - 18 | 2557 | 65 | |
XM_006714039.3 | XP_006714102.1 | 18 - 19 | 2818 | 65 | |
XM_011534385.2 | XP_011532687.1 | 17 - 18 | 2811 | 65 | |
XM_017008280.1 | XP_016863769.1 | 17 - 18 | 2988 | 65 | |
GenBank mRNA | AK316578.1 | - | 17 - 18 | 2575 | 65 |
M21574.1 | - | 17 - 18 | 2581 | 65 | |
M22734.1 | - | - | 2570 | 65 |