The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024524.3 | NP_078800.3 | 17 - 18 | 2249 | 74 |
XM_005269357.3 | XP_005269414.1 | 17 - 18 | 2270 | 74 | |
XM_011513120.2 | XP_011511422.1 | 18 - 19 | 2371 | 74 | |
XM_011513121.1 | XP_011511423.1 | 17 - 18 | 2249 | 74 | |
XM_011513122.1 | XP_011511424.1 | 17 - 18 | 1943 | 74 | |
XM_011513123.2 | XP_011511425.1 | 18 - 19 | 2376 | 74 | |
XM_011513124.2 | XP_011511426.1 | 18 - 19 | 2376 | 74 | |
GenBank mRNA | AJ306929.1 | - | 8 - 9 | 2773 | 74 |
AK024639.1 | - | 6 - 7 | 646 | 74 | |
AK225064.1 | - | 6 - 7 | 646 | 74 |