The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024524.3 | NP_078800.3 | 7 - 8 | 1034 | 135 |
XM_005269357.3 | XP_005269414.1 | 7 - 8 | 1055 | 135 | |
XM_011513120.2 | XP_011511422.1 | 8 - 9 | 1156 | 135 | |
XM_011513121.1 | XP_011511423.1 | 7 - 8 | 1034 | 135 | |
XM_011513123.2 | XP_011511425.1 | 8 - 9 | 1161 | 135 | |
XM_011513124.2 | XP_011511426.1 | 8 - 9 | 1161 | 135 |