Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GLS2
Entrez Gene ID
27165
Gene Name
glutaminase 2
Gene Aliases
GA, GLS, LGA, hLGA
Chromosome Location
Chr.12: 56470944 - 56488414 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001280796.1 NP_001267725.1 6 - 7 1071 73
NM_001280797.1 NP_001267726.1 5 - 6 941 73
NM_001280798.1 NP_001267727.1 3 - 4 719 73
NM_013267.3 NP_037399.2 6 - 7 1071 73
XM_005268797.1 XP_005268854.1 6 - 7 698 73
XM_017019180.1 XP_016874669.1 5 - 6 568 73
GenBank mRNA AF110330.1 - 6 - 7 73
AF223944.1 - 6 - 7 998 73
AK000467.1 - 6 - 7 818 73
AK294822.1 - 6 - 7 1016 73
AK303772.1 - 6 - 7 872 73
BC025661.1 - 5 - 6 656 73
DA060302.1 - 3 - 4 303 73
HM775422.1 - 5 - 6 690 73
HM775423.1 - 4 - 5 560 73
HM775424.1 - 5 - 6 690 73

View Full Product Details