Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ADAMTS19
Entrez Gene ID
171019
Gene Name
ADAM metallopeptidase with thrombospondin type 1 motif 19
Gene Aliases
-
Chromosome Location
Chr.5: 129460281 - 129738683 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_011543246.2 XP_011541548.1 14 - 15 1803 86
XM_011543247.2 XP_011541549.1 14 - 15 1827 86
XM_011543248.2 XP_011541550.1 14 - 15 1947 86
XM_011543249.2 XP_011541551.1 9 - 10 2106 86
XM_017009174.1 XP_016864663.1 13 - 14 2562 86
XM_017009175.1 XP_016864664.1 10 - 11 2859 86
XM_017009176.1 XP_016864665.1 15 - 16 1975 86
GenBank mRNA AJ311904.1 - 15 - 16 2403 86
AK122622.1 - 5 - 6 649 86
AK128729.1 - 13 - 14 1618 86

View Full Product Details