Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ADAMTS19
Entrez Gene ID
171019
Gene Name
ADAM metallopeptidase with thrombospondin type 1 motif 19
Gene Aliases
-
Chromosome Location
Chr.5: 129460281 - 129738683 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_011543246.2 XP_011541548.1 17 - 18 2230 136
XM_011543247.2 XP_011541549.1 17 - 18 2254 136
XM_011543248.2 XP_011541550.1 17 - 18 2374 136
XM_011543249.2 XP_011541551.1 12 - 13 2533 136
XM_017009174.1 XP_016864663.1 16 - 17 2989 136
XM_017009175.1 XP_016864664.1 13 - 14 3286 136
XM_017009176.1 XP_016864665.1 18 - 19 2402 136
GenBank mRNA AJ311904.1 - 18 - 19 2797 136
AK122622.1 - 8 - 9 1043 136

View Full Product Details