Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ADAMTS19
Entrez Gene ID
171019
Gene Name
ADAM metallopeptidase with thrombospondin type 1 motif 19
Gene Aliases
-
Chromosome Location
Chr.5: 129460281 - 129738683 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_011543246.2 XP_011541548.1 2 - 3 273 109
XM_011543247.2 XP_011541549.1 2 - 3 238 109
XM_011543248.2 XP_011541550.1 2 - 3 358 109
XM_017009176.1 XP_016864665.1 2 - 3 238 109
GenBank mRNA AJ311904.1 - 3 - 4 885 109
AK128729.1 - 2 - 3 238 109

View Full Product Details