Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ADAMTS19
Entrez Gene ID
171019
Gene Name
ADAM metallopeptidase with thrombospondin type 1 motif 19
Gene Aliases
-
Chromosome Location
Chr.5: 129460281 - 129738683 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_011543246.2 XP_011541548.1 6 - 7 737 117
XM_011543247.2 XP_011541549.1 6 - 7 761 117
XM_011543248.2 XP_011541550.1 6 - 7 881 117
XM_017009174.1 XP_016864663.1 5 - 6 1496 117
GenBank mRNA AJ311904.1 - 7 - 8 1349 117

View Full Product Details