The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_178468.5 | NP_848563.1 | 3 - 4 | 930 | 100 |
GenBank mRNA | AK091138.1 | - | 2 - 3 | 273 | 100 |
BC113483.1 | - | 3 - 4 | 909 | 100 |