The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001198626.1 | NP_001185555.1 | 3 - 4 | 619 | 95 |
NM_001198627.1 | NP_001185556.1 | 4 - 5 | 679 | 95 | |
NM_001198628.1 | NP_001185557.1 | 2 - 3 | 276 | 95 | |
NM_001198629.1 | NP_001185558.1 | 2 - 3 | 402 | 95 | |
NM_001198630.1 | NP_001185559.1 | 3 - 4 | 462 | 95 | |
NM_001198631.1 | NP_001185560.1 | 2 - 3 | 475 | 95 | |
NM_175634.2 | NP_783552.1 | 2 - 3 | 537 | 95 | |
XM_011517351.2 | XP_011515653.1 | 3 - 4 | 391 | 95 | |
XM_017013931.1 | XP_016869420.1 | 1 - 2 | 169 | 95 | |
GenBank mRNA | D14821.1 | - | 1 - 2 | 166 | 95 |
DA499596.1 | - | 3 - 4 | 392 | 95 | |
DA504975.1 | - | 3 - 4 | 499 | 95 | |
DA774453.1 | - | 2 - 3 | 375 | 95 | |
DA776968.1 | - | 3 - 4 | 474 | 95 | |
DA777240.1 | - | 2 - 3 | 361 | 95 | |
DA795825.1 | - | 4 - 5 | 495 | 95 | |
DA796892.1 | - | 2 - 3 | 276 | 95 |