The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000139.4 | NP_000130.1 | 4 - 5 | 483 | 153 |
XM_005273846.4 | XP_005273903.1 | 5 - 6 | 798 | 153 | |
XM_011544850.2 | XP_011543152.1 | 5 - 6 | 416 | 153 | |
XM_017017362.1 | XP_016872851.1 | 5 - 6 | 798 | 153 | |
GenBank mRNA | AK304487.1 | - | 4 - 5 | 483 | 153 |
AK310142.1 | - | 4 - 5 | 483 | 153 | |
BC074800.2 | - | 4 - 5 | 412 | 153 | |
BC074843.2 | - | 4 - 5 | 412 | 153 | |
D10583.1 | - | 4 - 5 | 431 | 153 | |
DA972757.1 | - | 4 - 5 | 483 | 153 | |
X66187.1 | - | 2 - 3 | 171 | 153 |