Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PICALM
Entrez Gene ID
8301
Gene Name
phosphatidylinositol binding clathrin assembly protein
Gene Aliases
CALM, CLTH, LAP
Chromosome Location
Chr.11: 85957171 - 86069881 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001008660.2 NP_001008660.1 14 - 15 1816 116
NM_001206946.1 NP_001193875.1 15 - 16 1945 116
NM_001206947.1 NP_001193876.1 14 - 15 1531 116
NM_007166.3 NP_009097.2 15 - 16 1966 116
XM_005274322.2 XP_005274379.1 15 - 16 1950 116
XM_005274323.2 XP_005274380.1 15 - 16 1935 116
XM_005274324.2 XP_005274381.1 15 - 16 1950 116
XM_005274325.2 XP_005274382.1 15 - 16 1935 116
XM_005274326.2 XP_005274383.1 15 - 16 1950 116
XM_005274327.2 XP_005274384.1 15 - 16 1950 116
XM_005274328.2 XP_005274385.1 15 - 16 1935 116
XM_005274329.3 XP_005274386.1 15 - 16 1935 116
XM_005274330.2 XP_005274387.1 14 - 15 1800 116
XM_005274331.2 XP_005274388.1 15 - 16 1950 116
XM_005274332.2 XP_005274389.1 14 - 15 1785 116
XM_005274333.2 XP_005274390.1 14 - 15 1800 116
XM_005274334.2 XP_005274391.1 15 - 16 1935 116
XM_005274335.2 XP_005274392.1 14 - 15 1785 116
XM_005274336.2 XP_005274393.1 14 - 15 1785 116
XM_005274337.2 XP_005274394.1 15 - 16 1950 116
XM_005274340.2 XP_005274397.1 14 - 15 1800 116
XM_006718699.2 XP_006718762.1 15 - 16 1929 116
XM_006718700.2 XP_006718763.1 15 - 16 1914 116
XM_006718701.3 XP_006718764.1 15 - 16 1929 116
XM_011545293.1 XP_011543595.1 13 - 14 1692 116
XM_017018381.1 XP_016873870.1 15 - 16 1914 116
XM_017018382.1 XP_016873871.1 14 - 15 1800 116
XM_017018383.1 XP_016873872.1 14 - 15 1800 116
XM_017018384.1 XP_016873873.1 14 - 15 1785 116
XM_017018385.1 XP_016873874.1 15 - 16 1935 116
XM_017018386.1 XP_016873875.1 14 - 15 1800 116
XM_017018387.1 XP_016873876.1 14 - 15 1785 116
XM_017018388.1 XP_016873877.1 13 - 14 1692 116
GenBank mRNA AB210017.1 - 15 - 16 1945 116
AB451308.1 - 14 - 15 1499 116
AK291414.1 - 16 - 17 1833 116
AK297062.1 - 6 - 7 686 116
AK300275.1 - 14 - 15 1508 116
BC048259.1 - 15 - 16 1727 116
BC064357.1 - 14 - 15 1778 116
BC073961.1 - 14 - 15 1652 116
U45976.1 - 15 - 16 1796 116

View Full Product Details