Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- PICALM
- Entrez Gene ID
-
8301
- Gene Name
- phosphatidylinositol binding clathrin assembly protein
- Gene Aliases
-
CALM, CLTH, LAP
- Chromosome Location
-
Chr.11: 85957171 - 86069881 on Build GRCh38
View Full Product Details