Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PICALM
Entrez Gene ID
8301
Gene Name
phosphatidylinositol binding clathrin assembly protein
Gene Aliases
CALM, CLTH, LAP
Chromosome Location
Chr.11: 85957171 - 86069881 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001008660.2 NP_001008660.1 2 - 3 585 93
NM_001206946.1 NP_001193875.1 2 - 3 585 93
NM_001206947.1 NP_001193876.1 2 - 3 300 93
NM_007166.3 NP_009097.2 2 - 3 585 93
XM_005274322.2 XP_005274379.1 2 - 3 569 93
XM_005274323.2 XP_005274380.1 2 - 3 569 93
XM_005274324.2 XP_005274381.1 2 - 3 569 93
XM_005274325.2 XP_005274382.1 2 - 3 569 93
XM_005274326.2 XP_005274383.1 2 - 3 569 93
XM_005274327.2 XP_005274384.1 2 - 3 569 93
XM_005274328.2 XP_005274385.1 2 - 3 569 93
XM_005274329.3 XP_005274386.1 2 - 3 569 93
XM_005274330.2 XP_005274387.1 2 - 3 569 93
XM_005274331.2 XP_005274388.1 2 - 3 569 93
XM_005274332.2 XP_005274389.1 2 - 3 569 93
XM_005274333.2 XP_005274390.1 2 - 3 569 93
XM_005274334.2 XP_005274391.1 2 - 3 569 93
XM_005274335.2 XP_005274392.1 2 - 3 569 93
XM_005274336.2 XP_005274393.1 2 - 3 569 93
XM_005274337.2 XP_005274394.1 2 - 3 569 93
XM_005274340.2 XP_005274397.1 2 - 3 569 93
XM_006718699.2 XP_006718762.1 2 - 3 569 93
XM_006718700.2 XP_006718763.1 2 - 3 569 93
XM_006718701.3 XP_006718764.1 2 - 3 569 93
XM_011545293.1 XP_011543595.1 2 - 3 569 93
XM_017018381.1 XP_016873870.1 2 - 3 569 93
XM_017018382.1 XP_016873871.1 2 - 3 569 93
XM_017018383.1 XP_016873872.1 2 - 3 569 93
XM_017018384.1 XP_016873873.1 2 - 3 569 93
XM_017018385.1 XP_016873874.1 2 - 3 569 93
XM_017018386.1 XP_016873875.1 2 - 3 569 93
XM_017018387.1 XP_016873876.1 2 - 3 569 93
XM_017018388.1 XP_016873877.1 2 - 3 569 93
GenBank mRNA AB210017.1 - 2 - 3 585 93
AB451308.1 - 2 - 3 268 93
AK291414.1 - 3 - 4 452 93
AK300275.1 - 2 - 3 277 93
BC048259.1 - 2 - 3 346 93
BC064357.1 - 2 - 3 547 93
BC073961.1 - 2 - 3 421 93
DC300074.1 - 2 - 3 300 93
U45976.1 - 2 - 3 415 93

View Full Product Details