Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PICALM
Entrez Gene ID
8301
Gene Name
phosphatidylinositol binding clathrin assembly protein
Gene Aliases
CALM, CLTH, LAP
Chromosome Location
Chr.11: 85957171 - 86069881 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001008660.2 NP_001008660.1 8 - 9 1120 128
NM_001206946.1 NP_001193875.1 8 - 9 1120 128
NM_001206947.1 NP_001193876.1 8 - 9 835 128
NM_007166.3 NP_009097.2 8 - 9 1120 128
XM_005274322.2 XP_005274379.1 8 - 9 1104 128
XM_005274323.2 XP_005274380.1 8 - 9 1104 128
XM_005274324.2 XP_005274381.1 8 - 9 1104 128
XM_005274325.2 XP_005274382.1 8 - 9 1104 128
XM_005274326.2 XP_005274383.1 8 - 9 1104 128
XM_005274327.2 XP_005274384.1 8 - 9 1104 128
XM_005274328.2 XP_005274385.1 8 - 9 1104 128
XM_005274329.3 XP_005274386.1 8 - 9 1104 128
XM_005274330.2 XP_005274387.1 8 - 9 1104 128
XM_005274331.2 XP_005274388.1 8 - 9 1104 128
XM_005274332.2 XP_005274389.1 8 - 9 1104 128
XM_005274333.2 XP_005274390.1 8 - 9 1104 128
XM_005274334.2 XP_005274391.1 8 - 9 1104 128
XM_005274335.2 XP_005274392.1 8 - 9 1104 128
XM_005274336.2 XP_005274393.1 8 - 9 1104 128
XM_005274337.2 XP_005274394.1 8 - 9 1104 128
XM_005274340.2 XP_005274397.1 8 - 9 1104 128
XM_006718699.2 XP_006718762.1 8 - 9 1104 128
XM_006718700.2 XP_006718763.1 8 - 9 1104 128
XM_006718701.3 XP_006718764.1 8 - 9 1104 128
XM_011545293.1 XP_011543595.1 8 - 9 1104 128
XM_017018381.1 XP_016873870.1 8 - 9 1104 128
XM_017018382.1 XP_016873871.1 8 - 9 1104 128
XM_017018383.1 XP_016873872.1 8 - 9 1104 128
XM_017018384.1 XP_016873873.1 8 - 9 1104 128
XM_017018385.1 XP_016873874.1 8 - 9 1104 128
XM_017018386.1 XP_016873875.1 8 - 9 1104 128
XM_017018387.1 XP_016873876.1 8 - 9 1104 128
XM_017018388.1 XP_016873877.1 8 - 9 1104 128
GenBank mRNA AB210017.1 - 8 - 9 1120 128
AB451308.1 - 8 - 9 803 128
AK291414.1 - 9 - 10 987 128
AK300275.1 - 8 - 9 812 128
BC048259.1 - 8 - 9 881 128
BC064357.1 - 8 - 9 1082 128
BC073961.1 - 8 - 9 956 128
U45976.1 - 8 - 9 950 128

View Full Product Details