The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_173593.3 | NP_775864.3 | 5 - 6 | 544 | 55 |
GenBank mRNA | AB089940.1 | - | 5 - 6 | 544 | 55 |
AK093503.1 | - | 3 - 4 | 266 | 55 | |
CN400443.1 | - | 5 - 6 | 451 | 55 |