Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PCM1
Entrez Gene ID
5108
Gene Name
pericentriolar material 1
Gene Aliases
PTC4, RET/PCM-1
Chromosome Location
Chr.8: 17922763 - 18029948 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001315507.1 NP_001302436.1 38 - 39 6444 100
NM_001315508.1 NP_001302437.1 35 - 36 5976 100
NM_006197.3 NP_006188.3 38 - 39 6468 100
XM_006716336.3 XP_006716399.1 41 - 42 6919 100
XM_006716340.3 XP_006716403.1 41 - 42 6907 100
XM_006716349.3 XP_006716412.1 37 - 38 6379 100
XM_011544528.2 XP_011542830.1 41 - 42 6940 100
XM_011544529.2 XP_011542831.1 43 - 44 7246 100
XM_011544530.2 XP_011542832.1 42 - 43 7062 100
XM_011544532.2 XP_011542834.1 41 - 42 6937 100
XM_011544533.2 XP_011542835.1 41 - 42 6928 100
XM_011544534.2 XP_011542836.1 41 - 42 6916 100
XM_011544535.2 XP_011542837.1 41 - 42 6904 100
XM_011544540.2 XP_011542842.1 41 - 42 6868 100
XM_017013472.1 XP_016868961.1 42 - 43 7056 100
XM_017013473.1 XP_016868962.1 42 - 43 7052 100
XM_017013474.1 XP_016868963.1 39 - 40 6670 100
XM_017013475.1 XP_016868964.1 41 - 42 6794 100
XM_017013476.1 XP_016868965.1 38 - 39 6378 100
XM_017013477.1 XP_016868966.1 41 - 42 6959 100
XM_017013478.1 XP_016868967.1 41 - 42 6937 100
XM_017013479.1 XP_016868968.1 41 - 42 6938 100
XM_017013480.1 XP_016868969.1 39 - 40 6610 100
XM_017013481.1 XP_016868970.1 41 - 42 6913 100
XM_017013482.1 XP_016868971.1 38 - 39 6574 100
XM_017013483.1 XP_016868972.1 40 - 41 6843 100
XM_017013484.1 XP_016868973.1 37 - 38 6269 100
XM_017013485.1 XP_016868974.1 40 - 41 6823 100
XM_017013486.1 XP_016868975.1 38 - 39 6505 100
XM_017013487.1 XP_016868976.1 38 - 39 6496 100
XM_017013488.1 XP_016868977.1 38 - 39 6493 100
XM_017013489.1 XP_016868978.1 40 - 41 6796 100
XM_017013490.1 XP_016868979.1 40 - 41 6772 100
XM_017013491.1 XP_016868980.1 37 - 38 6391 100
XM_017013492.1 XP_016868981.1 37 - 38 6388 100
XM_017013493.1 XP_016868982.1 39 - 40 6691 100
XM_017013494.1 XP_016868983.1 37 - 38 6376 100
XM_017013495.1 XP_016868984.1 39 - 40 6516 100
XM_017013496.1 XP_016868985.1 36 - 37 6099 100
XM_017013497.1 XP_016868986.1 37 - 38 6364 100
XM_017013498.1 XP_016868987.1 37 - 38 6355 100
XM_017013499.1 XP_016868988.1 39 - 40 6495 100
XM_017013500.1 XP_016868989.1 37 - 38 6352 100
XM_017013501.1 XP_016868990.1 39 - 40 6654 100
XM_017013502.1 XP_016868991.1 36 - 37 6075 100
GenBank mRNA AF068293.1 - 7 - 8 969 100
AI810388.1 - 1 - 2 100
AI924817.1 - 1 - 2 100
AK091406.1 - 15 - 16 2153 100
AK295856.1 - 16 - 17 2454 100
AK301727.1 - 15 - 16 1960 100
AK302378.1 - 17 - 18 2265 100
AW080255.1 - 1 - 2 100
BC133052.1 - 34 - 35 5626 100
BC140946.1 - 38 - 39 6273 100
BI966701.1 - 1 - 2 100
L27841.1 - 38 - 39 6455 100

View Full Product Details