Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GLE1
Entrez Gene ID
2733
Gene Name
GLE1, RNA export mediator
Gene Aliases
GLE1L, LCCS, LCCS1, hGLE1
Chromosome Location
Chr.9: 128504692 - 128542301 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001003722.1 NP_001003722.1 15 - 16 2137 93
XM_006717059.2 XP_006717122.1 17 - 18 2276 93
XM_006717060.2 XP_006717123.1 16 - 17 2249 93
XM_011518549.1 XP_011516851.1 17 - 18 2313 93
XM_011518550.1 XP_011516852.1 17 - 18 2164 93
XM_011518551.1 XP_011516853.1 16 - 17 2164 93
GenBank mRNA AK001767.1 - 10 - 11 1387 93
AK074801.1 - 14 - 15 1893 93
AK222854.1 - 15 - 16 2111 93
AK290533.1 - 14 - 15 1874 93
AL117584.1 - 13 - 14 1620 93
BC030012.1 - 15 - 16 2130 93

View Full Product Details