Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
A2ML1
Entrez Gene ID
144568
Gene Name
alpha-2-macroglobulin like 1
Gene Aliases
CPAMD9
Chromosome Location
Chr.12: 8822472 - 8887202 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_144670.5 NP_653271.2 3 - 4 583 81
XM_011520566.2 XP_011518868.1 3 - 4 501 81
XM_011520567.2 XP_011518869.1 3 - 4 501 81
XM_017018868.1 XP_016874357.1 3 - 4 501 81
XM_017018869.1 XP_016874358.1 3 - 4 501 81
XM_017018870.1 XP_016874359.1 3 - 4 501 81
GenBank mRNA AK122624.1 - 3 - 4 81
AK290355.1 - 3 - 4 436 81
AL832139.1 - 3 - 4 434 81
AL833479.1 - 1 - 2 99 81
BX648231.1 - 3 - 4 435 81
DA436610.1 - 3 - 4 501 81
DA456454.1 - 3 - 4 436 81

View Full Product Details