Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
A2ML1
Entrez Gene ID
144568
Gene Name
alpha-2-macroglobulin like 1
Gene Aliases
CPAMD9
Chromosome Location
Chr.12: 8822472 - 8887202 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_144670.5 NP_653271.2 6 - 7 827 77
XM_011520566.2 XP_011518868.1 6 - 7 745 77
XM_011520567.2 XP_011518869.1 6 - 7 745 77
XM_017018868.1 XP_016874357.1 6 - 7 745 77
XM_017018869.1 XP_016874358.1 6 - 7 745 77
XM_017018870.1 XP_016874359.1 6 - 7 745 77
GenBank mRNA AK122624.1 - 6 - 7 721 77
AK123601.1 - 3 - 4 204 77
AK290355.1 - 6 - 7 680 77
AL832139.1 - 6 - 7 678 77
AL833479.1 - 4 - 5 343 77
BX648231.1 - 6 - 7 679 77

View Full Product Details