The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006901.3 | NP_008832.2 | 14 - 15 | 2673 | 65 |
XM_006720539.2 | XP_006720602.1 | 14 - 15 | 2673 | 65 | |
XM_011521613.2 | XP_011519915.1 | 14 - 15 | 2673 | 65 | |
XM_011521614.2 | XP_011519916.1 | 15 - 16 | 2359 | 65 | |
XM_011521615.2 | XP_011519917.1 | 14 - 15 | 2526 | 65 | |
XM_011521616.2 | XP_011519918.1 | 14 - 15 | 2320 | 65 | |
XM_011521617.2 | XP_011519919.1 | 14 - 15 | 2673 | 65 | |
XM_011521618.2 | XP_011519920.1 | 14 - 15 | 2673 | 65 | |
XM_011521619.2 | XP_011519921.1 | 13 - 14 | 2616 | 65 | |
XM_011521620.2 | XP_011519922.1 | 13 - 14 | 2616 | 65 | |
XM_011521622.2 | XP_011519924.1 | 14 - 15 | 2673 | 65 | |
XM_017022228.1 | XP_016877717.1 | 14 - 15 | 2302 | 65 | |
XM_017022229.1 | XP_016877718.1 | 15 - 16 | 2508 | 65 | |
XM_017022230.1 | XP_016877719.1 | 13 - 14 | 2616 | 65 | |
GenBank mRNA | AB209841.1 | - | 12 - 13 | 2051 | 65 |
AB290183.1 | - | 13 - 14 | 2108 | 65 | |
AF117888.1 | - | 14 - 15 | 2424 | 65 | |
AJ001714.1 | - | 13 - 14 | 2289 | 65 | |
BC140869.1 | - | 14 - 15 | 2342 | 65 | |
BC150294.1 | - | 12 - 13 | 1606 | 65 |