The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006901.3 | NP_008832.2 | 20 - 21 | 3259 | 111 |
XM_006720539.2 | XP_006720602.1 | 20 - 21 | 3259 | 111 | |
XM_011521613.2 | XP_011519915.1 | 20 - 21 | 3259 | 111 | |
XM_011521614.2 | XP_011519916.1 | 21 - 22 | 2945 | 111 | |
XM_011521615.2 | XP_011519917.1 | 20 - 21 | 3112 | 111 | |
XM_011521616.2 | XP_011519918.1 | 20 - 21 | 2906 | 111 | |
XM_011521617.2 | XP_011519919.1 | 20 - 21 | 3259 | 111 | |
XM_011521618.2 | XP_011519920.1 | 20 - 21 | 3259 | 111 | |
XM_011521619.2 | XP_011519921.1 | 19 - 20 | 3202 | 111 | |
XM_011521620.2 | XP_011519922.1 | 19 - 20 | 3202 | 111 | |
XM_011521621.2 | XP_011519923.1 | 19 - 20 | 3139 | 111 | |
XM_011521622.2 | XP_011519924.1 | 20 - 21 | 3259 | 111 | |
XM_011521623.2 | XP_011519925.1 | 3 - 4 | 362 | 111 | |
XM_017022228.1 | XP_016877717.1 | 20 - 21 | 2888 | 111 | |
XM_017022229.1 | XP_016877718.1 | 21 - 22 | 3094 | 111 | |
XM_017022230.1 | XP_016877719.1 | 19 - 20 | 3202 | 111 | |
GenBank mRNA | AB209841.1 | - | 18 - 19 | 2637 | 111 |
AB290183.1 | - | 19 - 20 | 2694 | 111 | |
AF117888.1 | - | 20 - 21 | 3010 | 111 | |
BC140869.1 | - | 20 - 21 | 2928 | 111 | |
BC150294.1 | - | 18 - 19 | 2267 | 111 |