The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006901.3 | NP_008832.2 | 5 - 6 | 1590 | 89 |
XM_006720539.2 | XP_006720602.1 | 5 - 6 | 1590 | 89 | |
XM_011521613.2 | XP_011519915.1 | 5 - 6 | 1590 | 89 | |
XM_011521614.2 | XP_011519916.1 | 6 - 7 | 1276 | 89 | |
XM_011521615.2 | XP_011519917.1 | 5 - 6 | 1443 | 89 | |
XM_011521616.2 | XP_011519918.1 | 5 - 6 | 1237 | 89 | |
XM_011521617.2 | XP_011519919.1 | 5 - 6 | 1590 | 89 | |
XM_011521618.2 | XP_011519920.1 | 5 - 6 | 1590 | 89 | |
XM_011521621.2 | XP_011519923.1 | 5 - 6 | 1590 | 89 | |
XM_011521622.2 | XP_011519924.1 | 5 - 6 | 1590 | 89 | |
XM_017022229.1 | XP_016877718.1 | 6 - 7 | 1425 | 89 | |
GenBank mRNA | AB290183.1 | - | 4 - 5 | 1025 | 89 |
AF117888.1 | - | 5 - 6 | 1341 | 89 | |
AJ001714.1 | - | 4 - 5 | 1206 | 89 | |
BC060886.1 | - | 5 - 6 | 1556 | 89 | |
BC140869.1 | - | 5 - 6 | 1259 | 89 | |
DQ088983.1 | - | 3 - 4 | 227 | 89 |