Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
STX11
Entrez Gene ID
8676
Gene Name
syntaxin 11
Gene Aliases
FHL4, HLH4, HPLH4
Chromosome Location
Chr.6: 144140044 - 144191939 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_011536214.2 XP_011534516.1 1 - 2 366 128

View Full Product Details