The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001145940.1 | NP_001139412.1 | 1 - 2 | 101 | 65 |
XM_017026848.1 | XP_016882337.1 | 1 - 2 | 101 | 65 | |
GenBank mRNA | AA432316.1 | - | 1 - 2 | 64 |