Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
P2RX6
Entrez Gene ID
9127
Gene Name
purinergic receptor P2X 6
Gene Aliases
P2RXL1, P2X6, P2XM
Chromosome Location
Chr.22: 21009699 - 21028013 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001159554.1 NP_001153026.1 6 - 7 586 80
NM_005446.3 NP_005437.2 6 - 7 642 80
XM_005261819.3 XP_005261876.1 6 - 7 1366 80
XM_011530498.2 XP_011528800.1 6 - 7 1367 80
XM_011530499.2 XP_011528801.1 5 - 6 1296 80
XM_011530500.2 XP_011528802.1 6 - 7 1289 80
XM_011530501.2 XP_011528803.1 5 - 6 634 80
XM_011530502.2 XP_011528804.1 6 - 7 1367 80
XM_017029066.1 XP_016884555.1 6 - 7 1367 80
XM_017029067.1 XP_016884556.1 5 - 6 1294 80
XM_017029068.1 XP_016884557.1 6 - 7 1366 80
XM_017029069.1 XP_016884558.1 5 - 6 1215 80
XM_017029070.1 XP_016884559.1 5 - 6 568 80
XM_017029071.1 XP_016884560.1 5 - 6 1290 80
XM_017029072.1 XP_016884561.1 5 - 6 523 80
XM_017029073.1 XP_016884562.1 5 - 6 634 80
XM_017029074.1 XP_016884563.1 5 - 6 487 80
XM_017029075.1 XP_016884564.1 5 - 6 580 80
XM_017029076.1 XP_016884565.1 5 - 6 1278 80
GenBank mRNA AB002058.1 - 6 - 7 657 80
AF065385.1 - 6 - 7 656 80
BC033488.1 - 6 - 7 626 80
BC047287.1 - 6 - 7 625 80
BC063553.1 - 4 - 5 479 80
BC064805.1 - 4 - 5 438 80
BC109209.2 - 6 - 7 569 80
BI753477.1 - 4 - 5 438 80
CR456535.1 - 6 - 7 648 80

View Full Product Details