The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001317163.1 | NP_001304092.1 | 19 - 20 | 2773 | 79 |
NM_001317164.1 | NP_001304093.1 | 19 - 20 | 2444 | 79 | |
NM_001735.2 | NP_001726.2 | 19 - 20 | 2447 | 79 | |
XM_011518980.2 | XP_011517282.1 | 20 - 21 | 2816 | 79 | |
XM_017015102.1 | XP_016870591.1 | 21 - 22 | 2968 | 79 | |
XM_017015103.1 | XP_016870592.1 | 19 - 20 | 2601 | 79 | |
XM_017015104.1 | XP_016870593.1 | 21 - 22 | 2968 | 79 | |
GenBank mRNA | AB209031.1 | - | 19 - 20 | 2773 | 79 |
AK310774.1 | - | 19 - 20 | 2444 | 79 | |
BC113738.1 | - | 19 - 20 | 2442 | 79 | |
BC113740.1 | - | 19 - 20 | 2442 | 79 | |
M57729.1 | - | 19 - 20 | 2429 | 79 | |
M65134.1 | - | 9 - 10 | 1184 | 79 | |
T82068.1 | - | 2 - 3 | 163 | 79 |