Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C5
Entrez Gene ID
727
Gene Name
complement component 5
Gene Aliases
C5D, C5a, C5b, CPAMD4, ECLZB
Chromosome Location
Chr.9: 120952335 - 121075174 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001317163.1 NP_001304092.1 28 - 29 4017 114
NM_001735.2 NP_001726.2 28 - 29 3691 114
XM_011518980.2 XP_011517282.1 29 - 30 4060 114
XM_017015102.1 XP_016870591.1 30 - 31 4212 114
XM_017015103.1 XP_016870592.1 28 - 29 3845 114
GenBank mRNA AB209031.1 - 28 - 29 4015 114
BC113738.1 - 28 - 29 3686 114
BC113740.1 - 28 - 29 3686 114
M57729.1 - 28 - 29 3673 114
M65134.1 - 18 - 19 2428 114

View Full Product Details