The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006368.4 | NP_006359.3 | 3 - 4 | 798 | 97 |
GenBank mRNA | AF009368.1 | - | 3 - 4 | 358 | 97 |
AF029674.1 | - | 3 - 4 | 609 | 97 | |
AF211848.1 | - | 3 - 4 | 783 | 97 | |
AK299781.1 | - | 3 - 4 | 463 | 97 | |
AK313635.1 | - | 3 - 4 | 515 | 97 | |
BC009402.2 | - | 3 - 4 | 441 | 97 | |
BC010158.2 | - | 3 - 4 | 438 | 97 | |
FJ263669.1 | - | 3 - 4 | 345 | 97 | |
U88528.1 | - | 2 - 3 | 593 | 97 |