The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006368.4 | NP_006359.3 | 6 - 7 | 1068 | 63 |
GenBank mRNA | AF009368.1 | - | 6 - 7 | 628 | 63 |
AF029674.1 | - | 6 - 7 | 879 | 63 | |
AF211848.1 | - | 6 - 7 | 1053 | 63 | |
AK299781.1 | - | 6 - 7 | 733 | 63 | |
AK313635.1 | - | 6 - 7 | 785 | 63 | |
BC009402.2 | - | 6 - 7 | 711 | 63 | |
BC010158.2 | - | 6 - 7 | 708 | 63 | |
FJ263669.1 | - | 6 - 7 | 615 | 63 | |
U88528.1 | - | 5 - 6 | 863 | 63 |