The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001135024.1 | NP_001128496.1 | 5 - 6 | 830 | 124 |
NM_003477.2 | NP_003468.2 | 5 - 6 | 1159 | 124 | |
XM_011520390.1 | XP_011518692.1 | 5 - 6 | 583 | 124 | |
GenBank mRNA | AF001437.1 | - | 5 - 6 | 641 | 124 |
AK312463.1 | - | 5 - 6 | 655 | 124 | |
BC010389.1 | - | 5 - 6 | 663 | 124 | |
BI915296.1 | - | 5 - 6 | 506 | 123 | |
BX648910.1 | - | 5 - 6 | 717 | 124 | |
U79296.1 | - | 2 - 3 | 255 | 124 | |
U82328.1 | - | 5 - 6 | 671 | 124 | |
Y13145.1 | - | 5 - 6 | 633 | 124 |