The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000069.2 | NP_000060.2 | 11 - 12 | 1843 | 65 |
XM_005245478.3 | XP_005245535.1 | 11 - 12 | 1730 | 65 | |
GenBank mRNA | BC133671.1 | - | 11 - 12 | 1687 | 65 |
L33798.1 | - | 11 - 12 | 1841 | 65 |