The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000069.2 | NP_000060.2 | 34 - 35 | 4465 | 110 |
XM_005245478.3 | XP_005245535.1 | 33 - 34 | 4295 | 110 | |
GenBank mRNA | BC133671.1 | - | 34 - 35 | 4309 | 110 |