The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001323374.1 | NP_001310303.1 | 6 - 7 | 555 | 76 |
NM_001323375.1 | NP_001310304.1 | 6 - 7 | 587 | 76 | |
NM_001323376.1 | NP_001310305.1 | 3 - 4 | 557 | 76 | |
NM_001323377.1 | NP_001310306.1 | 4 - 5 | 315 | 76 | |
NM_001744.5 | NP_001735.1 | 5 - 6 | 703 | 76 | |
GenBank mRNA | AK311662.1 | - | 3 - 4 | 412 | 76 |
AK312840.1 | - | 5 - 6 | 558 | 76 | |
BC016695.1 | - | 5 - 6 | 547 | 76 | |
BC025687.1 | - | 5 - 6 | 545 | 76 | |
BG721313.1 | - | 5 - 6 | 562 | 76 | |
D30742.1 | - | 5 - 6 | 533 | 76 | |
L17000.1 | - | 5 - 6 | 497 | 76 | |
L24959.1 | - | 6 - 7 | 555 | 76 |