The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001323374.1 | NP_001310303.1 | 9 - 10 | 805 | 90 |
NM_001323375.1 | NP_001310304.1 | 9 - 10 | 837 | 90 | |
NM_001323376.1 | NP_001310305.1 | 6 - 7 | 807 | 90 | |
NM_001323377.1 | NP_001310306.1 | 7 - 8 | 565 | 90 | |
NM_001744.5 | NP_001735.1 | 8 - 9 | 953 | 90 | |
GenBank mRNA | AK311662.1 | - | 6 - 7 | 662 | 90 |
AK312840.1 | - | 8 - 9 | 808 | 90 | |
BC016695.1 | - | 8 - 9 | 797 | 90 | |
BC025687.1 | - | 8 - 9 | 795 | 90 | |
D30742.1 | - | 8 - 9 | 783 | 90 | |
L17000.1 | - | 8 - 9 | 747 | 90 | |
L24959.1 | - | 9 - 10 | 805 | 90 |