The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020139.3 | NP_064524.3 | 1 - 2 | 79 | 77 |
XM_005263140.3 | XP_005263197.1 | 1 - 2 | 74 | 77 | |
XM_006714274.3 | XP_006714337.1 | 1 - 2 | 74 | 77 | |
GenBank mRNA | AK291335.1 | - | 1 - 2 | 41 | 77 |
AY358841.1 | - | 1 - 2 | 45 | 77 | |
BC001953.1 | - | 1 - 2 | 72 | 77 | |
BC037277.2 | - | 1 - 2 | 41 | 77 | |
BC095414.1 | - | 1 - 2 | 47 | 77 | |
BI544872.1 | - | 1 - 2 | 100 | 77 |