Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
BDH2
Entrez Gene ID
56898
Gene Name
3-hydroxybutyrate dehydrogenase, type 2
Gene Aliases
DHRS6, EFA6R, PRO20933, SDR15C1, UCPA-OR, UNQ6308
Chromosome Location
Chr.4: 103077618 - 103099867 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_020139.3 NP_064524.3 9 - 10 784 82
XM_005263140.3 XP_005263197.1 8 - 9 665 82
XM_006714274.3 XP_006714337.1 10 - 11 821 82
XM_011532127.2 XP_011530429.1 10 - 11 863 82
XM_011532128.2 XP_011530430.1 9 - 10 821 82
XM_017008461.1 XP_016863950.1 10 - 11 860 82
XM_017008462.1 XP_016863951.1 9 - 10 818 82
GenBank mRNA AF164790.1 - 9 - 10 748 82
AK023323.1 - 8 - 9 1757 82
AK128463.1 - 9 - 10 2499 82
AK290160.1 - 9 - 10 740 82
AK300626.1 - 9 - 10 629 82
AY358841.1 - 9 - 10 750 82
BC001953.1 - 9 - 10 777 82
BC037277.2 - 8 - 9 1763 82

View Full Product Details