Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ANKRD35
Entrez Gene ID
148741
Gene Name
ankyrin repeat domain 35
Gene Aliases
-
Chromosome Location
Chr.1: 145866560 - 145885866 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001280799.1 NP_001267728.1 4 - 5 296 61
NM_144698.4 NP_653299.4 6 - 7 566 61
GenBank mRNA AK091120.1 - 6 - 7 531 61
AK294308.1 - 4 - 5 275 61
AK296861.1 - 5 - 6 473 61
BC101618.1 - 6 - 7 513 61
BC101620.1 - 6 - 7 513 61

View Full Product Details