The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014420.2 | NP_055235.1 | 2 - 3 | 373 | 57 |
XM_011544488.2 | XP_011542790.1 | 3 - 4 | 948 | 57 | |
XM_017013316.1 | XP_016868805.1 | 3 - 4 | 671 | 57 | |
GenBank mRNA | AF177397.1 | - | 2 - 3 | 379 | 57 |
AK301609.1 | - | 2 - 3 | 443 | 57 | |
BC107046.2 | - | 2 - 3 | 300 | 57 | |
BC107047.2 | - | 2 - 3 | 300 | 57 | |
BX105775.1 | - | 2 - 3 | 373 | 57 |