Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TFCP2L1
Entrez Gene ID
29842
Gene Name
transcription factor CP2-like 1
Gene Aliases
CRTR1, LBP-9, LBP9
Chromosome Location
Chr.2: 121216587 - 121285210 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_014553.2 NP_055368.1 2 - 3 309 73
XM_017003902.1 XP_016859391.1 2 - 3 317 73
XM_017003904.1 XP_016859393.1 2 - 3 317 73
XM_017003905.1 XP_016859394.1 2 - 3 317 73
GenBank mRNA AF198488.1 - 2 - 3 306 73
AL137740.1 - 2 - 3 310 73
BC064698.1 - 2 - 3 309 73

View Full Product Details