Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TFCP2L1
Entrez Gene ID
29842
Gene Name
transcription factor CP2-like 1
Gene Aliases
CRTR1, LBP-9, LBP9
Chromosome Location
Chr.2: 121216587 - 121285210 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_014553.2 NP_055368.1 8 - 9 950 100
XM_017003902.1 XP_016859391.1 8 - 9 958 100
XM_017003903.1 XP_016859392.1 3 - 4 257 100
GenBank mRNA AF198488.1 - 8 - 9 947 100
AL137740.1 - 8 - 9 949 100
BC064698.1 - 8 - 9 950 100

View Full Product Details