Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PHEX
Entrez Gene ID
5251
Gene Name
phosphate regulating endopeptidase homolog, X-linked
Gene Aliases
HPDR, HPDR1, HYP, HYP1, LXHR, PEX, XLH
Chromosome Location
Chr.X: 22032327 - 22251310 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000444.5 NP_000435.3 13 - 14 2042 95
NM_001282754.1 NP_001269683.1 13 - 14 2042 95
XM_011545533.1 XP_011543835.1 9 - 10 851 95
XM_011545536.2 XP_011543838.1 5 - 6 884 95
XM_017029579.1 XP_016885068.1 9 - 10 852 95
GenBank mRNA AD000712.1 - 13 - 14 1639 95
AK298030.1 - 13 - 14 1588 95
AK301528.1 - 12 - 13 1778 95
AK304551.1 - 6 - 7 879 95
BC105057.1 - 13 - 14 1564 95
BC105059.1 - 13 - 14 1564 95
U60475.1 - 13 - 14 1473 95
U75645.1 - 13 - 14 1556 95
U82970.1 - 13 - 14 2083 95
U87284.1 - 13 - 14 1681 95

View Full Product Details